Learn More
MilliporeSigma™ HybridSPE™ DPX Tip
Catalog No. 111009670 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Hamilton
INTEGRA
Tecan
Universal
Volume (Metric):
1 mL (Tip)
1.25 mL (Tip)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Volume (Metric)
11-100-9670 Hamilton 1 mL (Tip)
11-100-9714 Tecan 1 mL (Tip)
11-100-9716 Universal 1 mL (Tip)
11-100-9715 INTEGRA 1.25 mL (Tip)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-100-9670 Supplier MilliporeSigma™ Supplier No. 52978U
Add to Cart
Add to Cart
HybridSPE™ Phospholipid Removal Technology for Biological Matrices. Remove phospholipids and proteins for accurate and reproducible LC-MS analysis.
Key Features and Benefits:
  • More accurate results
  • Unique chemistry can effectively separate hydrophobic analytes (such as vitamins) from phospholipids, unlike competitive product chemistries
  • Elimination of proteins and phospholipid-induced ion suppression
  • Simultaneous removal of proteins and phospholipids
  • Simple, standardized methodology, analogous to traditional protein precipitation
  • Alternative to complex traditional SPE method development
  • Reproducible, consistent performance reduces need for reprocessing
  • Less instrument downtime and longer column life
  • Decreased run times by eliminating the need for gradients to clean columns between samples
  • High throughput processing that is automatable and compatible with common robotic systems
  • Ready-to-use, no preconditioning required
TRUSTED_SUSTAINABILITY
Specifications
Bed Weight 50 mg
For Use With (Application) Food and Beverage
For Use With (Equipment) Hamilton
Volume (Metric) 1 mL (Tip)