Learn More
BTX™ Hybrimune Chamber Cable
Catalog No. BTX470302 Shop All BTX Harvard Products
Change view
Click to view available options
Connection Type:
Chamber Cable
Catalog No. Connection Type
BTX470302 Chamber Cable
1 options
Catalog No. BTX470302 Supplier BTX™ Supplier No. 470302
Chamber Cable for BTX™ Hybrimune.
  • For connection to electrofusion generators
Specifications
For Use With (Application) In-Vivo Drug or Gene Delivery, Gene Therapy, Protein Delivery
For Use With (Equipment) Hybrimune
Product Type Cables & Adapters
Description Hybrimune Chamber Cable
Display LCD
Includes Cables Only
Sterility Non-sterile
Connection Type Chamber Cable
Interface Digital User
Pulse Form AC Sine Wave, DC Square Wave