Learn More
Electron Microscopy Sciences Hybrislip™ Thin Covers, 22 mm x 22 mm
SDP
Catalog No. 5026597
Encompass_Preferred
HybriSlips are flexible, ready-to-use thin (0.25 mm) covers for glass slides that minimize fluid friction and facilitate uniform reagent distribution. The RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Working surfaces are protected with clean release liners to prevent RNase contamination. Hybrislips remain flat and will not curl, even at high temperatures and are ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
Size: 22 mm x 22 mm
Thickness: 0.25 mm
Catalog No. 50-265-97 Supplier Electron Microscopy Sciences Supplier No. 7032922C
HybriSlips are flexible, ready-to-use thin (0.25 mm) covers for glass slides that minimize fluid friction and facilitate uniform reagent distribution. The RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Working surfaces are protected with clean release liners to prevent RNase contamination. Hybrislips remain flat and will not curl, even at high temperatures and are ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
Size: 22 mm x 22 mm
Thickness: 0.25 mm