Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Hybrislip™ Thin Covers, 24 mm x 30 mm
SDP

Supplier:  Electron Microscopy Sciences 7032930

Encompass_Preferred

HybriSlips are flexible, ready-to-use thin (0.25 mm) covers for glass slides that minimize fluid friction and facilitate uniform reagent distribution. The RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Working surfaces are protected with clean release liners to prevent RNase contamination. Hybrislips remain flat and will not curl, even at high temperatures and are ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
Size: 24 mm x 30 mm
Thickness: 0.25 mm

Catalog No. 50-266-00


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.