Learn More
Research Products International Corp HybriSlip Hybridization Covers, RNase Free, 24 x 60mm, 100 per Package
SDP
Catalog No. 50212538
Encompass
Hydrophobic covers will not trap or bind probes to their surfaces like glass coverslips. Allows for uniform distribution of probe reagent and prevent evaporation. HybriSlips remain flat and will not curl at high temperatures. Outperforms glass and films without added cost.
Catalog No. 50-212-538 Supplier Research Products International Corp Supplier No. 247457
Hydrophobic covers will not trap or bind probes to their surfaces like glass coverslips. Allows for uniform distribution of probe reagent and prevent evaporation. HybriSlips remain flat and will not curl at high temperatures. Outperforms glass and films without added cost.