Learn More
Research Products International Corp HybriWell Sealing Covers, 20mm Chamber Diameter, 30ul Chamber Volume, 100 per Package
SDP
Catalog No. 50212541
Encompass
HybriWell covers seal securely to a microscope slide surface in seconds and can be easily removed after heating. These RNase free, hydrophobic covers eliminate the need for silanizing agents. Access ports allow convenient addition and removal of sample.
Catalog No. 50-212-541 Supplier Research Products International Corp Supplier No. 247464
HybriWell covers seal securely to a microscope slide surface in seconds and can be easily removed after heating. These RNase free, hydrophobic covers eliminate the need for silanizing agents. Access ports allow convenient addition and removal of sample.