Learn More
Research Products International Corp HybriWell Sealing Covers, 22 x 40mm Chamber, 180-200ul Chamber Volume, 100 per Package
SDP
Catalog No. 50212543
Encompass
HybriWell covers seal securely to a microscope slide surface in seconds and can be easily removed after heating. These RNase free, hydrophobic covers eliminate the need for silanizing agents. Access ports allow convenient addition and removal of sample.
Catalog No. 50-212-543 Supplier Research Products International Corp Supplier No. 247467
HybriWell covers seal securely to a microscope slide surface in seconds and can be easily removed after heating. These RNase free, hydrophobic covers eliminate the need for silanizing agents. Access ports allow convenient addition and removal of sample.