Learn More
Research Products International Corp HybriWell Sealing Covers, 45 x 45mm Chamber, 150-300ul Chamber Volume, 50 per Package
SDP
Supplier:  Research Products International Corp 247468
Encompass
HybriWell covers seal securely to a microscope slide surface in seconds and can be easily removed after heating. These RNase free, hydrophobic covers eliminate the need for silanizing agents. Access ports allow convenient addition and removal of sample.
Catalog No. 50-197-7228