Learn More
Grace Bio Labs Inc HybriWell™ Sealing System, 100/pack
Catalog No. NC1827185
Encompass_Preferred
HBW6L2L | 6-9.8mm X 20mm X 0.25mm Depth | 50UL Approx. Vol., 25mm X 75mm OD | SecureSeal™ Adhesive Chamber | 1.5mm Diameter Ports | 200 Port Seals Included.
Catalog No. NC1827185 Supplier Grace Bio Labs Inc Supplier No. 612107
HBW6L2L | 6-9.8mm X 20mm X 0.25mm Depth | 50UL Approx. Vol., 25mm X 75mm OD | SecureSeal™ Adhesive Chamber | 1.5mm Diameter Ports | 200 Port Seals Included.