Learn More
MilliporeSigma Blank Strip Kit, Agar Strips, 50/pk
Catalog No. NC0465654
Encompass
Agar Strips are used for monitoring of ambient air and compressed gases with the RCS Air Samplers.
Catalog No. NC0465654 Supplier MilliporeSigma Supplier No. 1441070050
Agar Strips are used for monitoring of ambient air and compressed gases with the RCS Air Samplers.