Learn More
MilliporeSigma Cover Slides, 100/pk
Catalog No. NC1517145
Encompass
Cover slides covers the agar strip surface and prevents contamination post sampling and during incubation.
Catalog No. NC1517145 Supplier MilliporeSigma Supplier No. 1441110100
Cover slides covers the agar strip surface and prevents contamination post sampling and during incubation.