Learn More
Hycult Biotech Inc Hycult C5, Rat, mAb, clone 4G2 20ug
SDP
Catalog No. NC4034983
Encompass
Monoclonal antibody clone 4G2 recognizes human, rabbit and rat complement C5 and can be used for ELISA, Western blotting, and Functional Studies.
Catalog No. NC4034983 Supplier Hycult Biotech Inc Supplier No. HM304420UG
Monoclonal antibody clone 4G2 recognizes human, rabbit and rat complement C5 and can be used for ELISA, Western blotting, and Functional Studies.