Learn More
MSC Clorox Healthcare Cleaner Disinfectant, Hospital Grade, 32 fl oz, Spray Bottle
Catalog No. 502836518
Encompass
This Clorox Healthcare Hydrogen Peroxide Cleaner Disinfectant is a hospital-grade liquid disinfectant supplied in a 32 fl oz trigger spray bottle. It is formulated for effective cleaning and disinfection in healthcare environments, featuring EPA-approved claims against norovirus with a rapid 1-minute contact time.
  • Hospital grade disinfectant
  • Liquid form
  • Trigger spray bottle container
  • 32 fl oz capacity
  • Unscented formula
  • EPA-approved claims against norovirus (via accepted surrogate feline calicivirus)
  • 1-minute norovirus contact time
  • Safety data sheet available
  • Item is hazardous
Catalog No. 50-283-6518 Supplier MSC Supplier No. 65352981
This Clorox Healthcare Hydrogen Peroxide Cleaner Disinfectant is a hospital-grade liquid disinfectant supplied in a 32 fl oz trigger spray bottle. It is formulated for effective cleaning and disinfection in healthcare environments, featuring EPA-approved claims against norovirus with a rapid 1-minute contact time.
  • Hospital grade disinfectant
  • Liquid form
  • Trigger spray bottle container
  • 32 fl oz capacity
  • Unscented formula
  • EPA-approved claims against norovirus (via accepted surrogate feline calicivirus)
  • 1-minute norovirus contact time
  • Safety data sheet available
  • Item is hazardous