This technical summary highlights High Purity Products' Hydrogen Peroxide, a powerful oxidizing agent with a 35% concentration, available in a substantial 2320 lbs tote suitable for scientific and industrial applications. It is recognized for high purity, making it ideal for sensitive manufacturing environments. The product is environmentally friendly as it decomposes into water and oxygen without harmful residues. This packaging format optimizes large-scale operations and reduces the frequency of replenishment.
- High purity for demanding applications
- 35% concentration
- Clear, colorless liquid
- Tote packaging for large volume
- Decomposes into water and oxygen
- Used as an oxidizing agent, bleaching agent, antiseptic, and disinfectant
- Environmentally friendly with no harmful residues
- Storage recommendations: cool, dry place away from incompatible materials