Learn More
Grainger Chief Hydraulic Power Unit, 2.5 Gpm, 2300 Psi, 5 gal., 1 ea.
Catalog No. 502783354
Encompass_Preferred
This hydraulic power unit is designed for reliable fluid power applications, utilizing a 5 1/2 hp Honda engine and an integrated tank. It delivers a nominal flow rate of 2.5 gpm at a maximum pressure of 2300 psi for various industrial and mechanical operations.
  • Equipped with a 5 1/2 hp engine
  • Includes motor and tank assembly
  • Nominal flow rate of 2.5 gpm
  • Maximum pressure rating of 2300 psi
  • Relief valve range of 1500 psi to 3000 psi
  • Reservoir capacity of 5 gal.
  • Frame mount design
Catalog No. 50-278-3354 Supplier Grainger Supplier No. 19NE37
This hydraulic power unit is designed for reliable fluid power applications, utilizing a 5 1/2 hp Honda engine and an integrated tank. It delivers a nominal flow rate of 2.5 gpm at a maximum pressure of 2300 psi for various industrial and mechanical operations.
  • Equipped with a 5 1/2 hp engine
  • Includes motor and tank assembly
  • Nominal flow rate of 2.5 gpm
  • Maximum pressure rating of 2300 psi
  • Relief valve range of 1500 psi to 3000 psi
  • Reservoir capacity of 5 gal.
  • Frame mount design