Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydantoin, 99%

Catalog No. AAA1248636
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1248636 500 g
AAA1248622 100 g
AAA124860E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1248636 Supplier Thermo Scientific Chemicals Supplier No. A1248636
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 461-72-3 | C3H4N2O2 | 100.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-72-3
Molecular Formula C3H4N2O2
Molecular Weight (g/mol) 100.08
MDL Number MFCD00005259
InChI Key WJRBRSLFGCUECM-UHFFFAOYSA-N
Synonym hydantoin, 2,4-imidazolidinedione, glycolylurea, imidazole-2,4 3h,5h-dione, dantochlor, 2,4 3h,5h-imidazoledione, hydantoine, ccris 6532, 2-imidazolin-4 or 5-one, 2-hydroxy, 4h-imidazole-2,5-diol
PubChem CID 10006
ChEBI CHEBI:27612
IUPAC Name imidazolidine-2,4-dione
SMILES O=C1CNC(=O)N1

Specifications

Melting Point 221°C to 223°C
Quantity 500 g
Beilstein 110598
Merck Index 14,4761
Formula Weight 100.08
Percent Purity 99%
Chemical Name or Material Hydantoin
Hazard Category H373
Precautionary Statement P262-P314-P501c
EINECSNumber 207-313-3
RTECSNumber MT8210000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.