Learn More
Grainger Hydc Lift Kt w/Cstr 4 Post Elc 1000lb
Catalog No. 502767189
Encompass_Preferred
The Grainger Approved Hydc Lift Kt w/ Cstr 4 Post is a hydraulic lift kit with a 4 post design. It has a lifting capacity of 7,000 lbs and a maximum lift height of 72". The kit includes 4 columns, 4 adjustable arms, 2 hydraulic cylinders, and a power unit. It is designed for use with heavy duty vehicles and equipment. The lift features welded steel construction and a powder coat finish for durability. Safety features include an integrated locking system and hydraulic velocity fuses. This is an ideal lift kit for auto repair shops, fleet maintenance facilities, agriculture/construction companies, and other commercial operations.
Catalog No. 50-276-7189 Supplier Grainger Supplier No. 16Y689
Add to Cart
Add to Cart
The Grainger Approved Hydc Lift Kt w/ Cstr 4 Post is a hydraulic lift kit with a 4 post design. It has a lifting capacity of 7,000 lbs and a maximum lift height of 72". The kit includes 4 columns, 4 adjustable arms, 2 hydraulic cylinders, and a power unit. It is designed for use with heavy duty vehicles and equipment. The lift features welded steel construction and a powder coat finish for durability. Safety features include an integrated locking system and hydraulic velocity fuses. This is an ideal lift kit for auto repair shops, fleet maintenance facilities, agriculture/construction companies, and other commercial operations.