Learn More
Horizon Scientifc 20 Cu. Ft. Capacity Standard Manual Defrost Laboratory Freezer With Natural Refrigerants, 30 ¾ "W X 29” D X 74 5/8"H
SDP
Catalog No. NC1778262
Encompass_Preferred
The Standard Hydrocarbon Manual Defrost Freezers take a large step forward in performance from past solid door general purpose units; offering a microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. Models are ETL Listed and are backed by a two year parts and labor warranty, plus additional three year compressor parts warranty. :30 ¾ "W X 29” D X 74 5/8"HNOTE: THIS UNIT MUST HAVE 4” CLEARANCE ON SIDES AND BACK FOR ADEQUATE VENTILATION. Manual Defrost. 115V, 60 HZ, 6 AMPS, ½ HP
Catalog No. NC1778262 Supplier Horizon Scientifc Supplier No. ABTHCMFP20
The Standard Hydrocarbon Manual Defrost Freezers take a large step forward in performance from past solid door general purpose units; offering a microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies. Models are ETL Listed and are backed by a two year parts and labor warranty, plus additional three year compressor parts warranty. :30 ¾ "W X 29” D X 74 5/8"HNOTE: THIS UNIT MUST HAVE 4” CLEARANCE ON SIDES AND BACK FOR ADEQUATE VENTILATION. Manual Defrost. 115V, 60 HZ, 6 AMPS, ½ HP