Learn More
Grainger HYDE TOOLS Glass Cutter, 0.06 in Min Material
Catalog No. NC2606902
Encompass_Preferred
The Hyde Tools Glass Cutter is a handheld tool for scoring and cutting glass. It features a 0.06 inch minimum material thickness capacity and a tungsten carbide cutting wheel. The tool has an ergonomic soft grip handle and includes a breaker bar for snapping the glass along the score line. It can cut glass up to 1/8 inch thick and is designed for applications like glazing, stained glass work, and glass crafts. The cutter uses standard replacement wheels and comes with a protective sheath. It weighs 4 ounces and measures 6 inches long.
Catalog No. NC2606902 Supplier Grainger Supplier No. 36M387
The Hyde Tools Glass Cutter is a handheld tool for scoring and cutting glass. It features a 0.06 inch minimum material thickness capacity and a tungsten carbide cutting wheel. The tool has an ergonomic soft grip handle and includes a breaker bar for snapping the glass along the score line. It can cut glass up to 1/8 inch thick and is designed for applications like glazing, stained glass work, and glass crafts. The cutter uses standard replacement wheels and comes with a protective sheath. It weighs 4 ounces and measures 6 inches long.