Learn More
Thermco Products Inc Specific Gravity Hydrometers - Plain Form
Catalog No. NC1574905
Encompass_Preferred
Gravity Range: 1.000 to 1.220. 300mm Length.
Catalog No. NC1574905 Supplier Thermco Products Inc Supplier No. GW2540
Gravity Range: 1.000 to 1.220. 300mm Length.