Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution, For Diaphragmless cell, Chloroform-Free, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003011 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-011 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-003-011 Supplier Avantor Supplier No. 628501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Coulometric Karl Fischer Analysis

Specifications

Chemical Name or Material HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution
Name Note For Diaphragmless cell, Chloroform-Free
Quantity 500 mL
Flash Point 46.00°C
Packaging Narrow Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.