Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRA-POINT™ Titrant, 5mg/mL, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003172 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-003-172 2.5 L
02-003-171 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-003-172 Supplier Avantor Supplier No. 627468

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

    For Karl Fischer titration

Specifications

Chemical Name or Material HYDRA-POINT™ Titrant, 5mg/mL
Quantity 2.5 L
Flash Point 26.00°C
Packaging Narrow Mouth Amber Glass Bottle
Grade BAKER ANALYZED™ Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.