Learn More
Medchemexpress LLC Hydralazine-d4 hydrochloride | 2749234-32-8 | 1 MG
SDP
Catalog No. 5000405287
Encompass_Preferred
Hydralazine-d4 hydrochloride is the deuterium labeled form of Hydralazine hydrochloride, a direct-acting vasodilator used as an antihypertensive agent. This compound incorporates stable heavy isotopes for use as tracers in drug development, where deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-05287 Supplier Medchemexpress LLC Supplier No. HYB0464S1MG
Hydralazine-d4 hydrochloride is the deuterium labeled form of Hydralazine hydrochloride, a direct-acting vasodilator used as an antihypertensive agent. This compound incorporates stable heavy isotopes for use as tracers in drug development, where deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can serve as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS