Learn More
Medchemexpress LLC Hydralazine hydrochloride | 304-20-1 | 99.9% | 1 ML
SDP
Catalog No. 5000406431
Encompass_Preferred
Hydralazine hydrochloride is an antihypertensive agent that inhibits mitochondrial fission and human peritoneal mesothelial cell proliferation. It also possesses immunomodulation and anti-migratory effects, and activates the intrinsic pathway of apoptosis, causing DNA damage. This product is intended for research use only.
  • Inhibits mitochondrial fission
  • Inhibits human peritoneal mesothelial cell proliferation
  • Possesses immunomodulation and anti-migratory effects
  • Activates the intrinsic pathway of apoptosis
  • Causes DNA damage
  • Intended for research use only
Catalog No. 50-004-06431 Supplier Medchemexpress LLC Supplier No. HYB046410MM/1ML
Hydralazine hydrochloride is an antihypertensive agent that inhibits mitochondrial fission and human peritoneal mesothelial cell proliferation. It also possesses immunomodulation and anti-migratory effects, and activates the intrinsic pathway of apoptosis, causing DNA damage. This product is intended for research use only.
  • Inhibits mitochondrial fission
  • Inhibits human peritoneal mesothelial cell proliferation
  • Possesses immunomodulation and anti-migratory effects
  • Activates the intrinsic pathway of apoptosis
  • Causes DNA damage
  • Intended for research use only