Learn More
Abcam Hydralazine hydrochloride, DNA methylation inhibitor., 1G
Catalog No. 502707189
Encompass_Preferred
MW 196.64 g/mol, Purity >99%. Inhibits DNA methylation by inhibition of MAPK (EC₅₀ = 2.96 μM). Multiple, direct effects on vascular smooth muscle; hyperpolarization, inhibition of IP₃-induced release of Ca²+ and formation of NO leading to cGMP-mediated vasodilation. DNMT inhibitor (IC₅₀ = 2 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7189 Supplier Abcam Supplier No. AB1208631G
MW 196.64 g/mol, Purity >99%. Inhibits DNA methylation by inhibition of MAPK (EC₅₀ = 2.96 μM). Multiple, direct effects on vascular smooth muscle; hyperpolarization, inhibition of IP₃-induced release of Ca²+ and formation of NO leading to cGMP-mediated vasodilation. DNMT inhibitor (IC₅₀ = 2 μM).
The product is subject to the following: Abcam Restricted Use Statement