Learn More
AdipoGen Hydramethylnon
SDP
Supplier:  AdipoGen CDXH0096G001
Encompass_Preferred
Chemical. CAS 67485-29-4. Formula C25H24F6N4. MW 494.5. Synthetic. Insecticide primarily used against cockroaches and ants. Chemical class called trifluoromethyl aminohydrazone, which is a metabolic inhibitor. It works by inhibiting complex III in the mitochondrial inner membrane and leads to a halting of oxidative phosphorylation. Compound can be used as analytical reference material.
Catalog No. 50-256-0744