Learn More
HYDRANAL™ - Benzoic acid, Buffer substance for KF titration, Solstice
Shop All Solstice Research Chemicals ProductsDescription
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
Specifications
| Chemical Name or Material | Hydranal™-Benzoic Acid |
| CAS | 65-85-0 |
| Boiling Point | 249 - 250 °C |
| Physical Form | Powder |
| Quantity | 500 g |
| CAS Max % | 100.00 |
| Flash Point | 121°C (249.8°F) |
| Purity Grade Notes | Auxiliaries for KF titration |
| Packaging | Plastic bottle |
| Grade | Karl Fischer |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.