Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™-Buffer for Acids, Liquid buffer medium for Karl Fischer titration in acidic samples (based on imidazole), Solstice

Catalog No. 60090313 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-313 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-313 Supplier Solstice Supplier No. 348046X500MLUS/DEL

missing translation for 'validMainframeMsg'

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula O2S
Molecular Weight (g/mol) 64.06
InChI Key RAHZWNYVWXNFOC-UHFFFAOYSA-N
IUPAC Name (oxo-λ⁴-sulfanylidene)oxidane
SMILES O=S=O

Specifications

Boiling Point 65°C (1.013 hPa).The physical data is that of the main component.
CAS Min % ≥50.00, ≥30.00
CAS Max % <70.00, <50.00
CAS 67-56-1, 288-32-4, 7446-09-5
Density 0.960 g/cm3 (20°C)
Physical Form Liquid
Quantity 6 x 500 mL
Flash Point 11°C
Packaging Polycoated Bottle
Grade KF Titration
Hazard Category Danger
Hazard Statement H314+H225+H332+H360+H370
Precautionary Statement P280+P271+P270+P264+P260+P243+P242+P241+P240+P233+P210+P202+P201
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.