Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™

Catalog No. 60046477 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-477 6 x 500 mL
60-025-74 500 mL
60-025-73 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-046-477 Supplier Honeywell Chemicals Supplier No. 348276X500MLUS/DEL

Specifications

Chemical Name or Material HYDRANAL™ - Composite 1
Name Note Reagent for volumetric one-component Karl Fischer titration (methanol free)
Boiling Point 194°C
CAS Min % ≥5.0000%, ≥1.0000%, ≥5.0000%
CAS Max % <10.0000%, <5.0000%, <5.0000%, <10.0000%
CAS 288-32-4, 7553-56-2, 7446-09-5, 111-90-0, 68007-08-9
Density ∼1.11 g/mL
Quantity 6 x 500 mL
Flash Point 97°C
Packaging Glass Bottle
Synonym DE Solvent, 2-(2-Ethoxyethoxy)ethanol, CARBITOL™, Diethylene glycol ethyl ether, Ethyldiglycol
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.