Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™-Composite 2, Titrating agent for volumetric one-component Karl Fischer titration (methanol free), Solstice

Catalog No. 60090315 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-315 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-090-315 Supplier Solstice Supplier No. 348061LUS

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator's performance.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C3H5IN2
Molecular Weight (g/mol) 195.99
InChI Key JBOIAZWJIACNJF-UHFFFAOYSA-N
IUPAC Name 1H-imidazole hydroiodide
SMILES I.N1C=CN=C1

Specifications

Boiling Point No data available
CAS Min % ≥5.00, ≥70.00
CAS Max % <10.00, <5.00, <10.00
CAS 288-32-4, 7553-56-2, 111-90-0, 68007-08-9, 693-98-1
Density 1.110 g/cm3 (20°C)
Physical Form Liquid
Quantity 1 L
Flash Point 103°C
Packaging Polycoated Bottle
Grade One Component Volumetry
Hazard Category Danger
Hazard Statement H372+H360+H351
Precautionary Statement P201+P202+P260+P264+P270+P281
Recommended Storage Room Temperature
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.