Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ NEXTGEN, Solstice

Catalog No. 60049491 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 50 mL
6 x 500 mL
Chemical Name or Material:
NEXTGEN Coulomat A-FA
NEXTGEN Coulomat C-FA
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Chemical Name or Material
60-049-491 6 x 500 mL NEXTGEN Coulomat A-FA
60-049-492 6 x 50 mL NEXTGEN Coulomat C-FA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-049-491 Supplier Solstice Supplier No. 344716X500ML
Add to Cart
Add to Cart

Titrating agent for volumetric, one-component Karl Fischer titrations (Methanol-free)

Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NEXTGEN Coulomat A-FA
Physical Form Liquid
Quantity 6 x 500 mL
Packaging Glass Bottle
Solubility Information Completely Miscible in Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.