Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smart Chemical HYDRANAL™ - Composite 5, Solstice

Catalog No. 60048618 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-618 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Alternative option to part number 34805
Catalog No. 60-048-618 Supplier Solstice Supplier No. 348051LSC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alternative option to part number 34805

Titrating agent for volumetric one-component Karl Fischer titration (methanol free).

Solstice's Smart Chemical Hydranal™ bottles are embedded with an RFID chip, visible in the form of a new label, that captures essential information for every titration, including article code, product name, lot number, shelf life and numeric concentration.

  • Secure Data Transfer: Ensure correct and complete reagent data every time with traceable information flow.
  • Efficient System Setup: Reduce system set-up time with automated reagent data transfer.

The RFID label is currently compatible with Mettler Toledo's Karl Fischer Compact and Excellence line titrators (when upgraded with external RFID reader) and is available on our best-selling Hydranal Volumetric Titrants.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Composite 5
CAS 288-32-4, 68007-08-9, 7553-56-2, 111-90-0, 693-98-1
Density 1.17 g/cm3
Quantity 1 L
Flash Point 99°C
Packaging Mettler-Toledo Smart Chemical Bottle
Grade Karl Fischer
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.