Learn More
Description
Solstice's Smart Chemical Hydranal™ bottles are embedded with an RFID chip, visible in the form of a new label, that captures essential information for every titration, including article code, product name, lot number, shelf life and numeric concentration.
- Secure Data Transfer: Ensure correct and complete reagent data every time with traceable information flow.
- Efficient System Setup: Reduce system set-up time with automated reagent data transfer.
The RFID label is currently compatible with Mettler Toledo's Karl Fischer Compact and Excellence line titrators (when upgraded with external RFID reader) and is available on our best-selling Hydranal Volumetric Titrants.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Composite 5 K |
| CAS | 7446-09-5, 693-98-1, 288-32-4, 111-90-0, 10034-85-2 |
| Density | 1.15 g/mL |
| Quantity | 1 L |
| Flash Point | 90°C |
| Packaging | Mettler-Toledo Smart Chemical Bottle |
| Grade | Karl Fischer |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.