Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™-CompoSolver E, Honeywell Fluka™

Catalog No. 60046410 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-410 6 x 1 L
60-006-40 1 L
60-046-409 4 x 2.5 L
60-006-41 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-410 Supplier Honeywell Chemicals Supplier No. 347346X1L/DEL

Medium for volumetric one-component Karl Fischer titration (ethanol-based)

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-CompoSolver E
CAS 64-17-5, 693-98-1, 7446-09-5
Density 0.80g/mL
Quantity 6 x 1 L
Flash Point 12°C (53.6°F)
UN Number UN1170
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.