Learn More
HYDRANAL™ - Coulomat AF 7, For coulometer AF7, Solstice
Shop All Solstice Research Chemicals ProductsDescription
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™ - Coulomat AF 7 |
| Name Note | For coulometer AF7 |
| Boiling Point | 63°C |
| CAS Min % | ≥1.0000%, ≥5.0000% |
| CAS Max % | <10.0000%, <5.0000% |
| CAS | 288-32-4, 68007-08-9, 7446-09-5 |
| Density | 0.881 g/cm3 |
| Quantity | 6 x 1 L |
| Flash Point | 11°C |
| Packaging | Glass Bottle |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.