Learn More
Metrohm USA HYDRANAL-Coulomat AG, 1L
SDP
Supplier:  Metrohm USA SIGTI00626
Encompass
HYDRANAL-Coulomat AG. Reagent for coulometric KF titration (anolyte solution), for cells with/without diaphragm. Suitability: use tested, for cells with/without diaphragm, Reagent for coulometric KF titration (anolyte solution) 1L, Polycoated Bottle
Catalog No. NC0652126