Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Coulomat AK, Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm, Solstice
BIOPHARMAGSA_VA

Catalog No. 6000648 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-48 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-006-48 Supplier Solstice Supplier No. 348206X500MLUS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-86-4

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat AK
Name Note Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm
Boiling Point 60°C
CAS Min % ≥5.0000%, ≥1.0000%, ≥20.0000%
CAS Max % <20.0000%, <5.0000%, <5.0000%, <30.0000%
CAS 75-89-8, 68007-08-9, 67-66-3, 109-86-4, 7446-09-5
Density 1.200 g/cm3
Quantity 500 mL
Flash Point 32°C
Packaging Glass Bottle
UN Number UN1992
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.