Learn More
HYDRANAL™ - Coulomat CG-K, Reagent for coulometric KF titration in ketones (catholyte solution), Solstice
Shop All Solstice Research Chemicals Products

Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-Coulomat CG-K |
| CAS | 7446-09-5, 288-32-4, 123-39-7 |
| Packaging | Glass Bottle |
| Quantity | 10 x 5 mL Ampules |
| Flash Point | 64°C (147.2°F) |
| Grade | Coulometry |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.