Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Coulomat CG-K, Reagent for coulometric KF titration in ketones (catholyte solution), Solstice

Catalog No. 60050006 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Kit
10 x 5 mL Ampules
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-050-006 1 Kit
60-025-72 10 x 5 mL Ampules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-050-006 Supplier Solstice Supplier No. 348216X50ML/DEL

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat CG-K
Name Note Reagent for coulometric KF titration in ketones (catholyte solution)
Boiling Point 152°C
CAS Min % ≥1.0000%, ≥10.0000%
CAS Max % <5.0000%
CAS 288-32-4, 68007-08-9, 97-99-4, 123-39-7, 7446-09-5
Density 1.100 g/cm3
Quantity 1 Kit
Flash Point 64°C
Packaging Ampoule
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.