Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - CRM Water Standard 10.0, Certified reference material for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice

Catalog No. 60050001 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
10 x 8 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-050-001 10 x 8 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 60-050-001 Supplier Solstice Supplier No. 3442580ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified reference material for the calibration of Karl Fischer titrators.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - CRM Water Standard 10.0
Name Note Certified reference material for Karl Fischer titration (water content 10 mg/g = 1.0%)
Boiling Point 109°C
CAS Min % ≥20.0000%
CAS Max % <30.0000%, <70.0000%
CAS 71-36-3, 108-32-7
Density 1.000 g/cm3
Quantity 10 x 8 mL
Flash Point 32°C
Assay Percent Notes effective water volumetric
Molecular Formula C8H10
Packaging Ampoule
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1C
IUPAC Name 1,2-xylene
Molecular Weight (g/mol) 106.17
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.