Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Formamide dry, Solvent for KF titration, Solstice
BIOPHARMAGSA_VA

Catalog No. 6002552 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-025-52 1 L
60-090-265 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-025-52 Supplier Solstice Supplier No. 347246X1L
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Solvent for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Flash Point 175°C (347°F)
Density 1.134g/mL at 25°C
Quantity 1 L
CAS 75-12-7
Packaging Glass Bottle
Linear Formula HCONH2
UN Number NONH for all modes of transport
Beilstein 505995
Formula Weight 45.04g/mol
Chemical Name or Material HYDRANAL™-Formamide dry
Hazard Statement P201-P260-P280-P308 + P313
RTECSNumber LQ0525000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.