Learn More
HYDRANAL™ - LipoSolver MH, Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium), Solstice
Shop All Solstice Research Chemicals Products

Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-LipoSolver MH |
| Density | 0.80g/mL |
| CAS | 67-56-1, 111-27-3 |
| Quantity | 1 L |
| Flash Point | 11°C (51.8°F) |
| Packaging | Glass bottle |
| MDL Number | MFCD02687787 |
| UN Number | UN1230 |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.