Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Medium K, Solstice
BIOPHARMA

Catalog No. 6000658 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-58 6 x 1 L
60-090-317 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-006-58 Supplier Solstice Supplier No. 346986X1LUS
Add to Cart
Add to Cart
This item is not returnable. View return policy

Medium for volumetric one-component Karl Fischer titration in aldehydes and ketones (methanol free).

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Medium K
Boiling Point 58 °C
CAS Min % ≥30.00, ≥10.00
CAS Max % <30.00, <60.00
CAS 107-98-2, 75-89-8, 101023-55-6, 67-66-3
Density 1.25g/mL
Physical Form Liquid
Quantity 6 x 1 L
Flash Point 56°C (132.8°F)
Purity Grade Notes One component volumetry
Packaging Polycoated bottle
UN Number UN1992
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.