Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Methanol Rapid, Reagent for accelerated volumetric one-component KF titration (working medium), Solstice
BIOPHARMA

Catalog No. 6000662 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-62 2.5 L
60-006-61 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-006-62 Supplier Solstice Supplier No. 378174X2.5LUS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium for accelerated volumetric one-component Karl Fischer titration.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Methanol Rapid
CAS 67-56-1, 288-32-4, 7446-09-5
Packaging Poly-coated Bottle
Density 0.80g/mL
Quantity 2.5 L
Flash Point 11°C (51.8°F)
UN Number UN1230
Grade Karl Fischer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.