Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - NEXTGEN Coulomat AG-FI, Solstice

Catalog No. 60049490 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-049-490 6 x 500 mL
60-049-489 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-049-490 Supplier Solstice Supplier No. 344336X500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anolyte for coulometric Karl Fischer titrations, for cells with and without diaphragm

  • Methanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Coulomat AG-FI
Melting Point -98°C (Methanol)
CAS Min % ≥60%
Boiling Point 64°C to 65°C (Methanol)
Color Yellow
Density 0.893 g/cm3 (at 20°C)
Physical Form Liquid
CAS 67-56-1
Quantity 6 x 500 mL
CAS Max % ≤90%
Flash Point 14°C
Packaging Glass Bottle
UN Number UN1230
Solubility Information Completely Miscible in Water
Grade Karl Fischer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.