Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - NEXTGEN Solvent E-FI, Solstice

Catalog No. 60049488 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-049-488 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-049-488 Supplier Solstice Supplier No. 344316X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium for volumetric, two-component Karl Fischer titrations

  • Ethanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Solvent E-FI
Melting Point -115°C
CAS Min % ≥55%
Color Colorless
Density 0.895 g/cm3
pH 5 to 6
Physical Form Liquid
CAS 64-17-5
Quantity 6 x 1 L
CAS Max % ≤80%
Packaging Glass Bottle
UN Number UN1170
Solubility Information Completely Miscible in Water
Grade Karl Fischer
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.