Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - NEXTGEN Solvent FI, Solstice

Catalog No. 60049485 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-049-485 1 L
60-049-486 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-049-485 Supplier Solstice Supplier No. 344321L
Add to Cart
Add to Cart

Medium for volumetric, two-component Karl Fischer titrations

  • Methanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Solvent FI
CAS Min % ≥0.10%
Density 0.9 g/cm3 (at 20°C)
pH 5 to 6
Physical Form Liquid
CAS 147-85-3
Quantity 1 L
CAS Max % ≤5%
Flash Point 11°C
Packaging Glass Bottle
UN Number UN1230
Solubility Information Completely Miscible in Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.