Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Solstice
BIOPHARMA

Catalog No. 6000664 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-64 1 L
60-006-65 2.5 L
60-046-460 4 x 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-006-64 Supplier Solstice Supplier No. 348126X1LUS
Add to Cart
Add to Cart

Specifications

Chemical Name or Material HYDRANAL™-Solvent CM
CAS 288-32-4, 67-56-1, 67-66-3
Density 1.26g/mL
Quantity 1 L
Flash Point 100°C (212°F)
MDL Number MFCD00147198
UN Number UN2810
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.