Learn More
HYDRANAL™ - Solvent CM, Reagent for volumetric two-component KF titration in oils (working medium), Solstice
Shop All Honeywell Research Chemicals ProductsSpecifications
Specifications
| Chemical Name or Material | HYDRANAL™ - Solvent CM |
| Name Note | Reagent for volumetric two-component KF titration in oils (working medium) |
| CAS Min % | ≥1.0000%, ≥20.0000% |
| Boiling Point | 60°C to 65°C |
| CAS Max % | <5.0000%, <30.0000% |
| CAS | 288-32-4, 67-56-1, 7446-09-5 |
| Density | ∼1.26 g/mL |
| Quantity | 4 x 2.5 L |
| Flash Point | 100°C |
| MDL Number | MFCD00147198 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.