Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™
BIOPHARMA

Catalog No. p-7072130 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-70 1 L
60-006-71 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-006-70 Supplier Honeywell Chemicals Supplier No. 348006X1LUS
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Solvent
CAS 67-56-1, 288-32-4, 7446-09-5
Density 0.89g/mL
Quantity 1 L
Flash Point 11°C (51.8°F)
MDL Number MFCD00147197
UN Number UN3286
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.